Learn More
Blood Group A Mouse anti-Human, PE, Clone: NaM87-1F6, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565385
Catalog No. BDB565385
Add to cart
Description
Description
The NaM87-1F6 monoclonal antibody specifically binds to blood group A expressed on human erythrocytes. Clone NaM87-1F6 induces agglutination of human group A red blood cells. It does not react with human group B, or group O red blood cells.
Specifications
Specifications
Blood Group A
NaM87-1F6
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Mouse
Affinity Chromatography
RUO
Primary
Human
Flow Cytometry
PE
Affinity Purified
IgG3, κ
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal