Learn More
Mouse anti-BMP-8b, Clone: 1017, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243677
Catalog No. NBP243677
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot
BMP-8b Monoclonal antibody specifically detects BMP-8b in Human samples. It is validated for Western Blot.
Specifications
Specifications
BMP-8b
1017
Western Blot 1:1000-1:20000
Purified
BMP8B
Recombinant protein encompassing a sequence within the center region of human BMP8B. The exact sequence is proprietary.
Protein G purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot
Unconjugated
PBS with No Preservative
BMP-8, bone morphogenetic protein 8b, MGC131757
Mouse
IgG2a
0.1 ml
Cytokine Research
Primary
656