Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-c-Myc, Clone: MYC699, Azide Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Specifications

Antigen c-Myc
Clone MYC699
Concentration 1.0 mg/mL
Dilution Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, CyTOF-ready
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP23469202
SDP
View Documents
Novus Biologicals
NBP2346920.2 MG
0.2 mg Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

c-Myc Monoclonal antibody specifically detects c-Myc in Human, Primate, Mouse (Negative) samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Specifications

Specifications

c-Myc
1.0 mg/mL
Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF
Unconjugated
Mouse
Autophagy, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epitope Tags, Myc Epitope Tags, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
10mM PBS - BSA free with No Preservative
avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian)
MYC
IgG1 κ
Protein A or G purified
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
MYC699
Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, CyTOF-ready
Monoclonal
Purified
RUO
Human, Primate, Mouse (Negative)
P01106
4609
A synthetic peptide, corresponding to aa 408-439 (AEEQKLISEEDLLRKRREQLKHKLEQL-RNSCA) from C-terminus of human c-myc, coupled to KLH
Primary
Protein A purified
The c-Myc protein is a transcription factor, which is encoded by the c-Myc gene on human chromosome 8q24. c-Myc is commonly activated in a variety of tumor cells and plays an important role in cellular proliferation, differentiation, apoptosis and cell cycle progression. The phosphorylation of c-Myc has been investigated and previous studies have suggested a functional association between phosphorylation at Thr58/Ser62 by glycogen synthase kinase 3, cyclin dependent kinase, ERK2 and C-Jun N terminal Kinase (JNK) in cell proliferation and cell cycle regulation. Studies also have shown that c-Myc is essential for tumor cell development in vasculogenesis and angiogenesis that distribute blood throughout the cells, and which brought extensive attention in the development of new therapeutic approach for cancer treatment.
SDS
Documents

Documents

Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit