Learn More
C-peptide, Mouse, Clone: 5B8, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of C-peptide.
Manufacturer:  Abnova Corporation MAB2805
Catalog No. 89-135-242
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
C-peptide
5B8
Mouse monoclonal antibody raised against synthetic peptide of C-peptide.
In PBS, pH 7.4 (0.09% sodium azide)
Mouse
IgG1, kappa
RUO
Primary
This antibody bind with equal efficiency to the human free C-peptide and C-peptide region in proinsulin molecules.
ELISA, Radioimmune Assays
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide corresponding to human C-peptide.
200 ug
Store at 4°C.
Monoclonal
Human