Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nNOS/NOS Type I Mouse anti-Rat, Unlabeled, Clone: 52/nNOS/NOS Type I, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611853

Catalog No. BDB611853


Add to cart

Description

Description

Nitric oxide synthase (NOS) is a cell-type specific enzyme which catalyzes the synthesis of nitric oxide (NO). NO is a short-lived radical which transmits cellular signals involved in vasorelaxation, neurotransmission, and cytotoxicity. In neurons and endothelial cells, constitutive NOS (cNOS) is activated by agonists that increase intracellular Ca2+concentrations and enhance calmodulin binding. Neuronal NOS (nNOS or bNOS) and endothelial NOS (eNOS) have recognition sites for NADPH, FAD, FMN, and calmodulin and are regulated in a similar manner. However, both have been shown to be distinct gene products of about 155kDa and 140kDa, respectively, and the human forms show 52% amino acid identity to each other. nNOS and the 130 kDa inducible macrophage NOS (iNOS) share 51% amino acid homology. nNOS (neuronal NOS) is found in the cytoplasm of cell types such as neurons, skeletal muscle fibers, and lung epithelium. It is slightly larger than the other isoforms due to an N-terminal extension containing a PDZ domain through which it binds PSD-95 (post-synaptic density protein) and localizes to the cell membrane of synapses.

Specifications

Specifications

nNOS/NOS Type I
52/nNOS/NOS Type I
Unconjugated
Affinity Purified
Rat nNOS/NOS Type I a.a. 144-262
Affinity Chromatography
RUO
Primary
Rat
Western Blot, Western Blotting
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
150μg
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit