Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-C17orf53, Clone: 5F3H1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237406

Catalog No. NBP237406


Add to cart

Description

Description

C17orf53 Monoclonal antibody specifically detects C17orf53 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

C17orf53
5F3H1
Unconjugated
PBS with 0.05% Sodium Azide
chromosome 17 open reading frame 53, FLJ11594, hypothetical protein LOC78995, MGC3130
Mouse
IgG2a
Protein A or G purified
RUO
Primary
78995
Western Blot, ELISA
1 mg/mL
Western Blot 1:500 - 1:2000, ELISA 1:10000
Affinity Purified
C17orf53
Purified recombinant fragment of human C17ORF53 (AA: 282-527) expressed in E. Coli.
69.8 kDa
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only