Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C17orf58 Antibody (OTI6E9), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246593
Description
Ensure accurate, reproducible results in Western Blot
C17orf58 Monoclonal specifically detects C17orf58 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
C17orf58 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000 | |
chromosome 17 open reading frame 58, hypothetical protein LOC284018, MGC138278 | |
Mouse | |
11 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
OTI6E9 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
C17orf58 | |
Full length human recombinant protein of human C17orf58(NP_858041) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
284018 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction