Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calnexin Antibody (1C2.2D11), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP236570APC
Description
Calnexin Monoclonal specifically detects Calnexin in Human samples. It is validated for Flow (Intracellular).Specifications
Calnexin | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1C2.2D11 | |
Flow (Intracellular) | |
P27824 | |
CANX | |
Partial recombinant human Calnexin protein (between amino acids 1-300). [UniProt P27824] | |
0.1 mL | |
ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision | |
821 | |
Store at 4C in the dark. | |
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction