Learn More
Mouse anti-Calnexin, DyLight 405, Clone: 1C2.2D11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236570V
Catalog No. NBP236570V
Add to cart
Description
Description
Calnexin Monoclonal antibody specifically detects Calnexin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications
Specifications
Calnexin
1C2.2D11
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular)
P27824
calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Mouse
IgG2b Kappa
0.1 ml
ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision
Primary
821
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Purified
CANX
Partial recombinant human Calnexin protein (between amino acids 1-300). [UniProt P27824]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human