Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Calnexin, PerCP, Clone: 1C2.2D11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236570PCP

Catalog No. NBP236570P


Add to cart

Description

Description

Calnexin Monoclonal antibody specifically detects Calnexin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications

Specifications

Calnexin
1C2.2D11
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular)
P27824
calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Mouse
IgG2b Kappa
0.1 ml
ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision
Primary
821
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
PerCP
PBS with 0.05% Sodium Azide
Purified
CANX
Partial recombinant human Calnexin protein (between amino acids 1-300). [UniProt P27824]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit