Learn More
Carcinoembryonic, Mouse, Clone: 131-13881, Abnova™
Mouse monoclonal antibody raised against native Carcinoembryonic antigen.
Manufacturer:  Abnova Corporation MAB4036
Catalog No. 89-135-584
Add to cart
Description
Description
Specifications
Specifications
Carcinoembryonic
131-13881
Mouse monoclonal antibody raised against native Carcinoembryonic antigen.
In 10mM phosphate buffer, 150mM NaCl, pH 7.4 (0.09% sodium azide)
Mouse
IgG1, kappa
RUO
Primary
Reacts with carcinomas arising from the gastrointestinal site such as colon, stomach and small intestine. Reactivity may also be found in some breast, lung, or ovarian epithelial tumors. CEA is a tumor associated, complex glycoprotein with a molecular weight of 180 KDa.
ELISA
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Native purified Carcinoembryonic antigen from human colonic adenocarcinoma.
1 mg
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human