Learn More
Mouse anti-Caveolin-1 (p Tyr14), Clone: MM0680-4C29, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236783
Catalog No. NBP236783
Add to cart
Description
Description
Caveolin-1 p Tyr14 Monoclonal antibody specifically detects Caveolin-1 p Tyr14 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Caveolin-1 (p Tyr14)
MM0680-4C29
Unconjugated
A 0.2 um filtered solution in PBS with No Preservative
Purified
CAV1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Phosphopeptide Y14 site of human Caveolin-1.
Protein G purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
This antibody recognizes human phosphor-Y14 Caveolin-1 in direct ELISA and WB.
Human
Immunocytochemistry, Immunofluorescence, Western Blot
LYOPH
Western Blot 1:1000-2000, Immunocytochemistry/Immunofluorescence 1:50-200
Q03135
CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085
Mouse
IgG2
0.1 mg
Cancer, Caveolae Markers, Cellular Markers, Cholesterol Metabolism, Hypoxia, Lipid and Metabolism, Lipid Droplets, Signal Transduction
Primary
857
Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.