Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBL, Mouse, Clone: 3B12, Abnova™

Mouse monoclonal antibody raised against partial recombinant CBL.

Manufacturer:  Abnova Corporation MAB10513

Catalog No. 89-108-700


Add to cart

Description

Description

The cbl oncogene was first identified as part of a transforming retrovirus which induces mouse pre-B and pro-B cell lymphomas. As an adaptor protein for receptor protein-tyrosine kinases, it positively regulates receptor protein-tyrosine kinase ubiquitination in a manner dependent upon its variant SH2 and RING finger domains. Ubiquitination of receptor protein-tyrosine kinases terminates signaling by marking active receptors for degradation. [provided by RefSeq

Specifications

Specifications

CBL
3B12
Mouse monoclonal antibody raised against partial recombinant CBL.
In ascites (0.03% sodium azide)
Liquid
CBL
Recombinant protein corresponding to human CBL.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Murine, Rat
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user.
CBL
C-CBL/CBL2/RNF55
Mouse
IgG1
RUO
Primary
867
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit