Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-CCL16/HCC-4/LEC, Clone: LEC66, Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | CCL16/HCC-4/LEC |
---|---|
Clone | LEC66 |
Concentration | 1.0 mg/mL |
Dilution | ELISA 1-5ug/ml for coating, Functional |
Applications | ELISA, Functional Assay |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23466602
|
Novus Biologicals
NBP2346660.2 MG |
0.2 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
CCL16/HCC-4/LEC Monoclonal antibody specifically detects CCL16/HCC-4/LEC in Human samples. It is validated for ELISA, Functional.Specifications
CCL16/HCC-4/LEC | |
1.0 mg/mL | |
ELISA, Functional Assay | |
Unconjugated | |
Mouse | |
Human | |
O15467 | |
6360 | |
Recombinant human HCC-4 protein | |
Primary | |
Protein A purified | |
Shows chemotactic activity for lymphocytes and monocytes but not neutrophils. Also shows potent myelo-suppressive activity, suppresses proliferation of myeloid progenitor cells. Recombinant SCYA16 shows chemotactic activity for monocytes and THP-1 monocytes, but not for resting lymphocytes and neutrophils. Induces a calcium flux in THP-1 cells that were desensitized by prior expression to RANTES. Mainly expressed in liver, also found in spleen and thymus. Highly expressed in LPS- and IFN-gamma-activated monocytes, weakly in some lymphocytes, including natural killer cells, gamma-delta T-cells, and some T-cell clones. The ability of HCC-4 to markedly improve recognition of poorly immunogenic cells suggests that it could be an effective component of antitumor vaccines. |
LEC66 | |
ELISA 1-5ug/ml for coating, Functional | |
Monoclonal | |
Purified | |
RUO | |
10mM PBS - BSA free with No Preservative | |
C-C motif chemokine 16, chemokine (C-C motif) ligand 16, Chemokine CC-4, Chemokine LEC, CKb12, HCC-4Liver-expressed chemokine, LCC-1Lymphocyte and monocyte chemoattractant, LEC, liver CC chemokine-1, LMCSmall-inducible cytokine A16, monotactin-1, MTN-1, NCC4IL-10-inducible chemokine, NCC-4ILINCK, new CC chemokine 4, SCYA16MGC117051, SCYL4, small inducible cytokine subfamily A (Cys-Cys), member 16 | |
CCL16 | |
IgG1 κ | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
11 kDa |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title