Learn More
Mouse anti-CCL20/MIP-3 alpha, Clone: 206D9.05, HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals DDX0420HRPO0.05MG
Catalog No. DDX0420HRP5
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CCL20/MIP-3 alpha Monoclonal antibody specifically detects CCL20/MIP-3 alpha in Human samples. It is validated for ELISA, Immunohistochemistry-Frozen, Block/Neutralize.
Specifications
Specifications
CCL20/MIP-3 alpha
206D9.05
ELISA 1:100-1:2000, Immunohistochemistry-Frozen, Blocking/Neutralizing
NP_001123518
beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38
Mouse
IgG1
0.05 mg
Store at -20C in the dark. Avoid freeze-thaw cycles.
Monoclonal
Human
ELISA, Immunohistochemistry (Frozen), Blocking Assay
HRP
Antibody in Tris-NaCl (pH 8.0) with No Preservative
Purified
CCL20
MIP-3 Alpha/CCL20 was used as immunogen.
Ion exchange chromatography
RUO
Primary
6364