Learn More
Mouse anti-CCR3, Clone: 5E8-G9-B4, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP1433370.1MG
 View more versions of this product
Catalog No. NBP14333701
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CCR3
5E8-G9-B4
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR-3, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841
Mouse
IgG2b Kappa
0.1 mg
Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary
1232
Flow Cytometry
0.5 mg/mL
Flow Cytometry 1:10-1:1000
Affinity Purified
CCR3
The immunogen for this antibody was CCR3.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human