Learn More
CD106 Mouse anti-Human, BUV737, Clone: 51-10C9, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565418
Catalog No. BDB565418
Add to cart
Description
Description
The 51-10C9 monoclonal antibody specifically binds to CD106. CD106 is a 100-110 kDa type I transmembrane sialoglycoprotrein that is also known as Vascular cell adhesion molecule-1 (VCAM-1) and INCAM-110. CD106 is expressed at high levels on the surface of cytokine-stimulated endothelium, and at minimal levels on unstimulated endothelium. VCAM-1 serves as a ligand for the leukocyte integrins α 4β 1 (CD49d/CD29 complex; VLA-4) and α 4β 7 (LPAM-1). The 51-10C9 monoclonal antibody inhibits the in vitro binding of lymphocytes and monocytes to VCAM-1 on stimulated endothelium.
Due to spectral differences between labeled cells and beads, using BD™ CompBeads can result in incorrect spillover values when used with BD Horizon BUV737 reagents. Therefore, the use of BD CompBeads or BD CompBeads Plus to determine spillover values for these reagents is not recommended. Different BUV737 reagents (e.g., CD4 vs. CD45) can have slightly different fluorescence spillover therefore, it may also be necessary to use clone specific compensation controls when using these reagents.
Specifications
Specifications
CD106
51-10C9
VCAM-1; Vascular cell adhesion protein 1; INCAM-100; L1CAM
Affinity Purified
IgG1, κ
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Brilliant Ultraviolet 737
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
Affinity Chromatography
RUO
Primary
Human