Learn More
CD107b Mouse anti-Human, Alexa Fluor 647, Clone: H4B4, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565305
Catalog No. BDB565305
Add to cart
Description
Description
The H4B4 monoclonal antibody specifically binds to CD107b. CD107b is a heavily glycosylated, ~120 kDa type I transmembrane protein that is also known as lysosomal-associated membrane protein 2 (LAMP-2/LAMP2), LAMPB, or LGP110. CD107b is expressed in the lysosomal and endosomal membranes of most cell types where it may play a role in maintaining membrane integrity. CD107b is also expressed on the surface of activated platelets and lymphocytes, including T lymphocytes, e.g., degranulating CD8+ cytotoxic T cells. CD107b serves as a ligand for selectins, including E-selectin, and mediates cellular adhesion. CD107a/ LAMP-1 and CD107b /LAMP-2 are carriers for poly-N-acetyllactosamines, and display the sialyl Le[x] antigen. CD107b is likewise expressed on some tumor cells and cell lines, including U937 and KG1a, and may facilitate tumor metastases.
Specifications
Specifications
CD107b
H4B4
LAMP-2; LAMP2; LAMPB; LGP110
Affinity Purified
Human Adult Adherent Peripheral Blood Cells
Affinity Chromatography
RUO
Primary
Human
Flow Cytometry, Immunocytochemistry
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal