Learn More
CD107b Mouse anti-Human, Brilliant Violet 786, Clone: H4B4, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565304
Catalog No. BDB565304
Add to cart
Description
Description
The H4B4 monoclonal antibody specifically binds to CD107b. CD107b is a heavily glycosylated, ∽120 kDa type I transmembrane protein that is also known as lysosomal-associated membrane protein 2 (LAMP-2/LAMP2), LAMPB, or LGP110. CD107b is expressed in the lysosomal and endosomal membranes of most cell types where it may play a role in maintaining membrane integrity. CD107b is also expressed on the surface of activated platelets and lymphocytes, including T lymphocytes, e.g., degranulating CD8+ cytotoxic T cells. CD107b serves as a ligand for selectins, including E-selectin, and mediates cellular adhesion. CD107a/ LAMP-1 and CD107b /LAMP-2 are carriers for poly-N-acetyllactosamines, and display the sialyl Le[x] antigen. CD107b is likewise expressed on some tumor cells and cell lines, including U937 and KG1a, and may facilitate tumor metastases.
The antibody was conjugated to BD Horizon BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).
Specifications
Specifications
CD107b
H4B4
LAMP-2; LAMP2; LAMPB; LGP110
Affinity Purified
Human Adult Adherent Peripheral Blood Cells
Affinity Chromatography
RUO
Primary
Human
Flow Cytometry, Immunocytochemistry
Brilliant Violet 786
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgG1, κ
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal