Learn More
CD11a Mouse anti-Human, APC-R700, Clone: HI111, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565771
Catalog No. BDB565771
Add to cart
Description
Description
The HI111 monoclonal antibody specifically binds to CD11a, the 180 kDa integrin α chain. This type I transmembrane glycoprotein associates with CD18 (integrin β 2) to form the heterodimeric glycoprotein CD11a/CD18. This heterodimer is also known as the lymphocyte (leukocytes) function associated antigen-1 (LFA-1) that is expressed on all leukocytes. LFA-1 is an adhesion molecule involved in lymphocyte and granulocyte functions. LFA-1 mediates adhesion of lymphoid cells to the vascular endothelium in association with its ligand, and the intracellular adhesion molecule-1 (ICAM-1), CD54. Other ligands are ICAM-2 (CD102) and ICAM-3 (CD50).
Specifications
Specifications
CD11a
HI111
LFA-1α; Lymphocyte (Leukocyte) function-associated antigen 1 α chain; ITGAL
Affinity Purified
IgG1, κ
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
APC-R700
Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide.
Murine
Affinity Chromatography
RUO
Primary
Baboon, Cynomolgus, Human, Rhesus