Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-CD163, APC, Clone: 6E10.1G6, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236494APC

Catalog No. NBP236494J

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CD163
6E10.1G6
Flow Cytometry
Purified
CD163
A synthetic peptide made to the human CD163 protein sequence (between residues 200-400). [Swiss-Prot Q86VB7]
125.5 kDa
0.1 ml
Immune System Diseases, Immunology, Innate Immunity
Primary
9332
Human, Mouse
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130
Mouse
IgG2b Kappa
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This antibody is specific for the extracellular domain of human CD163 protein
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit