Learn More
Mouse anti-CD163, Clone: 6E10.1G6, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236494
 View more versions of this product
Catalog No. NBP236494
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD163
6E10.1G6
Unconjugated
PBS with 0.05% Sodium Azide
CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130
Mouse
IgG2b Kappa
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
This antibody is specific for the extracellular domain of human CD163 protein
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
1.0 mg/mL
Western Blot 2 ug/ml, Flow Cytometry 2.5 ug/million cells, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500, Flow (Cell Surface)
Purified
CD163
A synthetic peptide made to the human CD163 protein sequence (between residues 200-400). [Swiss-Prot Q86VB7]
125.5 kDa
0.1 mg
Immune System Diseases, Immunology, Innate Immunity
Primary
9332
Human, Mouse