Learn More
Mouse anti-CD163, DyLight 650, Clone: 6E10.1G6, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236494C
Catalog No. NBP236494C
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD163
6E10.1G6
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
CD163
A synthetic peptide made to the human CD163 protein sequence (between residues 200-400). [Swiss-Prot Q86VB7]
125.5 kDa
0.1 ml
Immune System Diseases, Immunology, Innate Immunity
Primary
9332
Human, Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130
Mouse
IgG2b Kappa
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This antibody is specific for the extracellular domain of human CD163 protein