Learn More
Mouse anti-CD19, Clone: HIB19, PE-Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP14296725TESTS
 View more versions of this product
Catalog No. NBP14296725
Add to cart
Description
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
CD19
HIB19
PE-Cyanine7
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Mouse
IgG1 Kappa
25 Tests
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primary
930
Flow Cytometry
0.2mg/mL
Flow Cytometry 1:10-1:1000
Affinity Purified
CD19
The immunogen for this antibody was CD19.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human