Learn More
CD195 Mouse anti-Human, Brilliant Violet 650, Clone: 3A9, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564999
Catalog No. BDB564999
Add to cart
Description
Description
The 3A9 monoclonal antibody recognizes CD195, which is also known as the chemokine receptor, CCR5, a seven transmembrane-spanning G protein-associated molecule. The 3A9 antibody also reportedly cross-reacts with human CCR8. Results of epitope mapping and sequence comparison between CCR5 and CCR8 reveals that the first three amino acid residues for these two receptors are identical: MDY (Met-Asp-Tyr). CCR5 belongs to the β-chemokine recept or family. It is expressed on subsets of T lymphocytes, NK cells, monocytes, macrophages, and dendritic cells . CCR5 regulates lymphocyte chemotaxis activation and transendothelial migration during inflammation. It signals a response to at least three chemokines: RANTES and macrophage inflammatory protein-1 (MIP-1) α and β. Additionally, CCR5 has been found to be a co-receptor for macrophage-tropic HIV-1 on CD4+ cells, a characteristic that is important in viral transmission. Reports indicate that individuals who have partial (heterozygous) or complete (homozygous) deletion of the CCR5 allele, demonstrate resistance to HIV infection. CCR5 has been clustered as CD195 in the VIIth HLDA workshop.
Specifications
Specifications
CD195
3A9
CCR-5; Chemokine (C-C motif) receptor 5; CMKBR5; CKR5; CKR-5; CHEMR13
Affinity Purified
Human CCR5 Transfected Cell Line
Affinity Chromatography
RUO
Primary
Cynomolgus, Human, Rhesus Monkey
Flow Cytometry
Brilliant Violet 650
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgG2a, κ
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal