Learn More
CD1b Mouse anti-Human, Clone: SN13, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP14345925UG
 View more versions of this product
Catalog No. NB018553
Add to cart
Description
Description
CD1b Monoclonal antibody specifically detects CD1b in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
Specifications
CD1b
SN13
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Affinity Purified
CD1B
The immunogen for this antibody was CD1b.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human
Immunoprecipitation, Immunohistochemistry (Frozen), Flow Cytometry, Immunohistochemistry
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
CD1, CD1A, CD1b antigen, CD1B antigen, b polypeptide, CD1b molecule, cortical thymocyte antigen CD1B, differentiation antigen CD1-alpha-3, MGC125990, MGC125991, R1, T-cell surface glycoprotein CD1b
Mouse
IgG1
25 ug
Immunology
Primary
910