Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1d Antibody (51.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP1434600.1MG
Description
CD1d Monoclonal specifically detects CD1d in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
CD1d | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
antigen-presenting glycoprotein CD1d, CD1A, CD1d antigen, CD1D antigen, d polypeptide, CD1d molecule, differentiation antigen CD1-alpha-3, HMC class I antigen-like glycoprotein CD1D, MGC34622, R3, R3G1, T-cell surface glycoprotein CD1d, thymocyte antigen CD1D | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG2b |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
51.1 | |
Unconjugated | |
PBS, pH 7.2 with 0.09% Sodium Azide | |
CD1D | |
The immunogen for this antibody was CD1d. | |
0.1 mg | |
B Cell Development and Differentiation Markers | |
912 | |
Store at 4C. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction