Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-CD20, Clone: 2H7, PE,
Mouse Monoclonal Antibody
$219.36 - $1698.75
Specifications
| Antigen | CD20 |
|---|---|
| Clone | 2H7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABF1635
|
MilliporeSigma™
MABF1635 |
25 Tests |
Each for $219.36
|
|
|||||
|
MABF1636
|
MilliporeSigma™
MABF1636 |
100 Tests |
Each for $456.56
|
|
|||||
|
MABF1637
|
MilliporeSigma™
MABF1637 |
500 Tests |
Each for $1,698.75
|
|
|||||
Description
Specifically detects CD20 clone: 2H7 in Human samples, and it is validated for Flow Cytometry
The 2H7 antibody reacts with human CD20, expressed by pre-B cells, resting and activated B cells, follicular dendritic cells and a small subset of T cells, but not on plasma cells. CD20 is a 33-37kDa transmembrane protein which is differentially phosphorylated in resting and active B cells. The CD20 molecule plays a role in regulating B-cell activation.The 2H7 antibody is reported to be cross-reactive with several non-human species including Baboon, Chimpanzee, Cynomolgus and Rhesus.Specifications
| CD20 | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Inflammation and Immunology | |
| Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
| MS4A1; CD20 | |
| Primary | |
| Stable for one year at 2°C to 8°C from date of receipt. Note: It is recommended to store the product undiluted at 2°C to 8°C and protected from prolonged exposure to light. Do not freeze. |
| 2H7 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P11836 | |
| Immunogen information for this antibody is proprietary information. | |
| Purified by size-exclusion chromatography |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title