Learn More
CD22, Mouse, Clone: MEM-01, Abnova™
Mouse monoclonal antibody raised against native CD22.
Manufacturer:  Abnova Corporation MAB0908
Catalog No. 89-108-236
Add to cart
Description
Description
Specifications
Specifications
CD22
MEM-01
Mouse monoclonal antibody raised against native CD22.
In PBS, pH 7.4 (0.09% sodium azide)
Liquid
CD22
Native purified CD22 from Raji human Burkitt's lymphoma cell line.
100 ug
Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Primate
Flow Cytometry, Immunoprecipitation, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
CD22
FLJ22814/MGC130020/SIGLEC-2/SIGLEC2
Mouse
IgG1
RUO
Primary
933