Learn More
Mouse anti-CD23/Fc epsilon RII, Clone: EBVCS2, PE-Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP14296625TESTS
 View more versions of this product
Catalog No. NBP14296625
Add to cart
Description
Description
CD23/Fc epsilon RII Monoclonal antibody specifically detects CD23/Fc epsilon RII in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD23/Fc epsilon RII
EBVCS2
Flow Cytometry 1:10-1:1000
Affinity Purified
FCER2
The immunogen for this antibody was CD23.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE-Cyanine7
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor
Mouse
IgG1
25 Tests
Adaptive Immunity, Cancer, Immunology
Primary
2208