Learn More
Mouse anti-CD25/IL-2 R alpha, Clone: BC96, PE-Cy5, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP143093100TESTS
 View more versions of this product
Catalog No. NBP143093
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD25/IL-2 R alpha
BC96
PE-Cyanine5
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Mouse
IgG1 Kappa
100 Tests
Adaptive Immunity, Apoptosis, Cytokine Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction
Primary
3559
Flow Cytometry
0.2 mg/mL
Flow Cytometry 1:10-1:1000
Affinity Purified
IL2RA
The immunogen for this antibody was CD25.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human