Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD266 (TWEAK-R) Mouse anti-Human, PE, Clone: ITEM-1, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 565731

Catalog No. BDB565731


Add to cart

Description

Description

The ITEM-1 monoclonal antibody specifically binds to CD266, which is also known as TWEAK Receptor (TWEAK-R/TWEAKR), or Fibroblast growth factor inducible 14 (FGF-inducible 14, FN14). CD266 is a ~14 kDa type I transmembrane protein that is the 12A family member of the tumor necrosis factor receptor superfamily (TNFRSF12A). It is expressed at low levels in a variety of tissues including, heart, placenta, lung, muscle, and pancreas. CD266 is relatively highly expressed on HUVEC cells and certain tumor cell lines. TWEAK-induced CD266-mediated signaling can play roles in inflammation, the induction of apoptosis in certain cell types, the proliferation and migration of endothelial cells, and can promote angiogenesis within healthy and diseased tissues, eg, tumors. The ITEM-1 antibody can reportedly induce the proliferation of HUVEC cells and the death of certain TWEAK-sensitive tumor target cell lines. CD266 binds TWEAK (CD255) and can signal intracellularly by interactions with TRAF1, 2, and 3 cytoplasmic proteins leading to NF- κ B activation.

Specifications

Specifications

CD266 (TWEAK-R)
ITEM-1
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
TWEAKR; TWEAK-R; TWEAK Receptor; TNFRSF12A; FGF-inducible 14; FN14
Affinity Purified
Human
Affinity Chromatography
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit