Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-CD27/TNFRSF7, Clone: 203.6, Novus Biologicals™

Mouse Monoclonal Antibody

$129.00 - $299.00

Specifications

Antigen CD27/TNFRSF7
Clone 203.6
Concentration 0.2 mg/mL
Immunogen Sezary cells
Gene ID (Entrez) 939
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP24457401 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2445740.1MG
0.1 mg Each for $299.00
Add to cart
 
NBP24457400 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2445740.02MG
0.02 mg Each for $129.00
Add to cart
 
NBP24457402 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2445740.2MG
0.2 mg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence

CD27/TNFRSF7 Monoclonal antibody specifically detects CD27/TNFRSF7 in Human samples. It is validated for Flow Cytometry, Immunofluorescence.
Specifications

Specifications

CD27/TNFRSF7
0.2 mg/mL
939
Monoclonal
Mouse
IgG3 Kappa
Purified
RUO
Protein A or G purified
203.6
Sezary cells
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Primary
Recognizes a protein of a disulfide-linked 120kDa dimer, identified as CD27 (Workshop VI; Code 6T-028). CD27 is expressed on the majority of peripheral T cells, medullary thymocytes, memory-type B cells, and natural killer cells. It is a transmembrane phosphoglycoprotein that belongs to the tumor necrosis factor receptor (TNFR) superfamily. CD27 binds to its ligand CD70, a member of the TNF family, and induces T-cell co-stimulation and B-cell activation. It also interacts with TRAFs and is involved in activation of NFB and SAPK/JNK and induces apoptosis.
Unconjugated
Flow Cytometry, Immunofluorescence
Store at 4C.

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit