Learn More
Mouse anti-CD27/TNFRSF7, Clone: O323, PE-Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142962100TESTS
 View more versions of this product
Catalog No. NBP142962
Add to cart
Description
Description
CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD27/TNFRSF7
O323
PE-Cyanine7
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7
Mouse
IgG1 Kappa
100 Tests
Apoptosis, Cancer, Immunology, Innate Immunity
Primary
939
Flow Cytometry
0.2 mg/mL
Flow Cytometry 1:10-1:1000
Affinity Purified
CD27
The immunogen for this antibody was CD27.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human