Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD38 Antibody (FS02), Novus Biologicals™
Mouse Monoclonal Antibody
$277.00 - $518.50
Specifications
Antigen | CD38 |
---|---|
Clone | FS02 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml |
Applications | Flow Cytometry, SDS-Page, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24456500
|
Novus Biologicals
NBP2445650.02MG |
0.02 mg |
Each for $277.00
|
|
|||||
NBP24456501
|
Novus Biologicals
NBP2445650.1MG |
0.1 mg |
Each for $518.50
|
|
|||||
NBP24456502
|
Novus Biologicals
NBP2445650.2MG |
0.2 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
CD38 Monoclonal specifically detects CD38 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunofluorescence.Specifications
CD38 | |
0.2 mg/mL | |
Flow Cytometry, SDS-Page, Immunofluorescence | |
Unconjugated | |
Mouse | |
Apoptosis, Cell Biology, Hematopoietic Stem Cell Markers, Immunology, Stem Cell Markers | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
952 | |
Human CD38 | |
Primary | |
Store at 4C. |
FS02 | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10 | |
CD38 | |
IgG1 κ | |
Protein A or G purified | |
This antibody reacts with a type II membrane glycopeptide with a transmembrane sequence near the NH2-terminus. CD38 is a type II transmembrane glycoprotein that is present on early B- and T-cell lineages and activated B- and T-cells but is absent from most mature resting peripheral lymphocytes. CD38 is also found on thymocytes, pre-B cells, germinal center B-cells, mitogen-activated T-cells, monocytes and Ig-secreting plasma cells. CD38 is expressed on CD34+ cells. The CD34+CD38- population of hematopoietic stems cells defines the most pluripotent cells (e.g. blast colony forming cells). |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title