Learn More
Mouse anti-CD40 Ligand/TNFSF5, Clone: LL48, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals DDX0140P1000.05MG
Catalog No. DDX0140P05
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD40 Ligand/TNFSF5 Monoclonal antibody specifically detects CD40 Ligand/TNFSF5 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional, Neutralization, Block/Neutralize.
Specifications
Specifications
CD40 Ligand/TNFSF5
LL48
Unconjugated
Tris-NaCl (pH 8.0) with No Preservative
CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40-L, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5IMD3, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5
Mouse
IgG2a
0.05 mg
Adaptive Immunity, Apoptosis, Asthma, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors
Primary
959
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay, Blocking Assay, Neutralization
0.5 mg/mL
Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Functional, Blocking/Neutralizing 50-200 molar excess, Neutralization
Purified
CD40LG
CD40L/CD154 was used as immunogen.
Ion exchange chromatography
RUO
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Human