Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length recombinant CD40.

Manufacturer:  Abnova Corporation H00000958A01

Catalog No. 89-104-230


Add to cart

Description

Description

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor has been found to be essential in mediating a broad variety of immune and inflammatory responses including T cell-dependent immunoglobulin class switching, memory B cell development, and germinal center formation. AT-hook transcription factor AKNA is reported to coordinately regulate the expression of this receptor and its ligand, which may be important for homotypic cell interactions. Adaptor protein TNFR2 interacts with this receptor and serves as a mediator of the signal transduction. The interaction of this receptor and its ligand is found to be necessary for amyloid-beta-induced microglial activation, and thus is thought to be an early event in Alzheimer disease pathogenesis. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq

Sequence: MVRLPLQCVLWGCLLTAVHPEPPTACREKQYLINSQCCSLCQPGQKLVSDCTEFTETECLPCGESEFLDTWNRETHFHQHKYCDPNLGLRVQQKGTSETDTICTCEEGWHCTSEACESCVLHRSCSPGFGVKQIDICQPHFPKDRGLNLLM
Specifications

Specifications

CD40
Unconjugated
50% glycerol
BC064518
Bp50/CDW40/MGC9013/TNFRSF5/p50
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA, Western Blot
Mouse polyclonal antibody raised against a full-length recombinant CD40.
CD40
Antisera
CD40
CD40 (AAH64518, 1 a.a. to 151 a.a) full-length recombinant protein with GST tag.
RUO
Primary
958
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only