Learn More
Mouse anti-CD45RA, Clone: HI100, Cy5.5, PerCP, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP14285025TESTS
 View more versions of this product
Catalog No. NBP14285025
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD45RA
HI100
PerCP-Cyanine5.5
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Mouse
IgG2b Kappa
25 Tests
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary
5788
Flow Cytometry
0.2 mg/mL
Flow Cytometry 1:10-1:1000
Affinity Purified
PTPRC
The immunogen for this antibody was CD45RA.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human