Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD48/SLAMF2 Antibody (156-4H9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
$204.00 - $482.50
Specifications
Antigen | CD48/SLAMF2 |
---|---|
Clone | 156-4H9 |
Concentration | 0.5 mg/mL |
Dilution | Flow Cytometry 1:10-1:1000 |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14340303
|
Novus Biologicals
NBP1434030.025MG |
0.025 mg |
Each for $204.00
|
|
|||||
NBP14340301
|
Novus Biologicals
NBP1434030.1MG |
0.1 mg |
Each for $482.50
|
|
|||||
Description
CD48/SLAMF2 Monoclonal specifically detects CD48/SLAMF2 in Human samples. It is validated for Flow Cytometry.Specifications
CD48/SLAMF2 | |
0.5 mg/mL | |
Flow Cytometry | |
Unconjugated | |
RUO | |
Human | |
BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2 | |
CD48 | |
IgG1 | |
Affinity Purified |
156-4H9 | |
Flow Cytometry 1:10-1:1000 | |
Monoclonal | |
Mouse | |
Cell Biology, Cellular Markers, Immunology, Stem Cell Markers | |
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
962 | |
The immunogen for this antibody was CD48. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title