Learn More
Mouse anti-CD5, Clone: UCHT2, PE-Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142972100TESTS
 View more versions of this product
Catalog No. NBP142972
Add to cart
Description
Description
CD5 Monoclonal antibody specifically detects CD5 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD5
UCHT2
Flow Cytometry 1:10-1:1000
Affinity Purified
CD5
The immunogen for this antibody was CD5.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
This antibody is reactive to Human, and recognizes Primate CD 5.
Flow Cytometry
PE-Cyanine7
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1
Mouse
IgG1 Kappa
100 Tests
Adaptive Immunity, Cell Biology, Cytokine Research, Immunology
Primary
921
Human