Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD62L Mouse anti-Human, BB515, Clone: DREG-56, BD

Mouse Monoclonal Antibody

$105.00 - $220.00

Specifications

Antigen CD62L
Isotype IgG1, κ
Immunogen Supernatant from PMA-activated Human Peripheral Blood Leukocytes
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Conjugate BB515
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB564743
View Documents
BD Biosciences
564743
25 Tests Each for $105.00
Add to cart
 
BDB564742
View Documents
BD Biosciences
564742
100 Tests Each for $220.00
Add to cart
 
Description

Description

The DREG-56 monoclonal antibody specifically binds to CD62L. CD62L is a 76-95 kDa glycoprotein that is also referred to as L-selectin or LECAM-1. CD62L is expressed on neutrophils, monocytes, T- and B-lymphocyte subsets and NK cells. The DREG-56 antibody recognizes the same antigen as LAM-1, and specifically inhibits >90% of binding of human lymphocytes to high endothelial venules (HEV) in frozen sections of peripheral, but not mucosal lymphoid tissue. It thus defines L-selectin as a human lymphocyte homing receptor for peripheral lymph node HEV.

The antibody was conjugated to BD Horizon™ BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (eg, 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

Specifications

Specifications

CD62L
Supernatant from PMA-activated Human Peripheral Blood Leukocytes
BB515
Flow Cytometry
RUO
DREG-56
Affinity Chromatography
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
IgG1, κ
Aqueous buffered solution containing ≤0.09% sodium azide.
Primary
Murine
Affinity Purified
Monoclonal
SELL; L-selectin; LSEL; LAM-1; LECAM-1; LEU8; LNHR; MEL-14; PLNHR; TQ-1

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit