Learn More
CD62L Mouse anti-Human, PE-Cy7, Clone: DREG-56, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565535
Catalog No. BDB565535
Add to cart
Description
Description
The DREG-56 monoclonal antibody specifically binds to CD62L. CD62L is a 76-95 kDa glycoprotein that is also referred to as L-selectin or LECAM-1. CD62L is expressed on neutrophils, monocytes, T- and B-lymphocyte subsets and NK cells. The DREG-56 antibody recognizes the same antigen as LAM-1, and specifically inhibits >90% of binding of human lymphocytes to high endothelial venules (HEV) in frozen sections of peripheral, but not mucosal lymphoid tissue. It thus defines L-selectin as a human lymphocyte homing receptor for peripheral lymph node HEV.
Specifications
Specifications
CD62L
DREG-56
SELL; L-selectin; LSEL; LAM-1; LECAM-1; LEU8; LNHR; MEL-14; PLNHR; TQ-1
Affinity Purified
Supernatant from PMA-activated Human Peripheral Blood Leukocytes
Affinity Chromatography
RUO
Primary
Human
Flow Cytometry
PE-Cyanine7
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal