Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-CD63, Clone: SPM524, Azide Free, Novus Biologicals™

Mouse Monoclonal Antibody

$366.50

Specifications

Antigen CD63
Clone SPM524
Concentration 1.0 mg/mL
Immunogen Recombinant human CD63 protein
Gene Accession No. P08962
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP23477901
View Documents
Novus Biologicals
NBP2347790.1 MG
0.1 mg
Each for $366.50
Add to cart
 
NBP23477902
View Documents
Novus Biologicals
NBP2347790.2 MG
0.2 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

CD63 Monoclonal antibody specifically detects CD63 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

CD63
1.0 mg/mL
P08962
10mM PBS - BSA free with No Preservative
This product is specific for CD63
Primary
Mouse
Unconjugated
Protein A purified
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
SPM524
Recombinant human CD63 protein
967
Protein A purified
Monoclonal
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1 Kappa
Purified
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit