Learn More
Mouse anti-CD68/SR-D1, Clone: KP1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP2445390.02MG
 View more versions of this product
Catalog No. NBP24453900
Add to cart
Description
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
CD68/SR-D1 Monoclonal antibody specifically detects CD68/SR-D1 in Human, Feline, Monkey, Canine (Negative), Chicken (Negative), Porcine (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin, SDS-Page, Flow (Intracellular), Immunofluorescence.
Specifications
Specifications
CD68/SR-D1
KP1
Unconjugated
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1
Mouse
IgG1 Kappa
Protein A or G purified
RUO
Store at 4C.
Monoclonal
This antibody recognizes a glycoprotein of 110kDa, which is identified as CD68. It is important for identifying macrophages in tissue sections. It stains macrophages in a wide variety of human tissues, including Kupffer cells and macrophages in the red pulp of the spleen, in lamina propria of the gut, in lung alveoli, and in bone marrow. It reacts with myeloid precursors and peripheral blood granulocytes. It also reacts with plasmacytoid T cells, which are supposed to be of monocyte/macrophage origin. It shows strong granular cytoplasmic staining of chronic and acute myeloid leukemia and also reacts with rare cases of true histiocytic neoplasia. Lymphomas are negative or show few granules.
Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence
0.2mg/mL
Flow Cytometry 5 - 10 ul/million cells in 0.1ml, Immunohistochemistry-Paraffin 1:100-1:200, SDS-Page, Immunofluorescence 1:50 - 1:100
Purified
CD68
Subcellular fraction of human alveolar macrophages
110 kDa
0.02 mg
Cell Biology, Cytokine Research, Immunology
Primary
968
Human, Feline, Monkey, Canine (Negative), Chicken (Negative), Porcine (Negative)