Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-CD7, Clone: B-F12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP2447990.02MG

 View more versions of this product

Catalog No. NBP24479900


Add to cart

Description

Description

Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence

CD7 Monoclonal antibody specifically detects CD7 in Human, Porcine, Guinea Pig samples. It is validated for Flow Cytometry, SDS-Page, Immunofluorescence.
Specifications

Specifications

CD7
B-F12
Unconjugated
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein
Mouse
IgG2a Kappa
Protein A or G purified
RUO
Store at 4C.
Monoclonal
Recognizes a protein of 40kDa, identified as CD7 (also known as gp40, Leu9). CD7 is a member of the immunoglobulin gene superfamily. Its N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxyl-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. It is also found on natural killer cells, a small subpopulation of normal B cells and on malignant B cells. Cross-linking surface CD7 positively modulates T cell and NK cell activity as measured by calcium fluxes, expression of adhesion molecules, cytokine secretion and proliferation. CD7 associates directly with phosphoinositol 3'-kinase. CD7 ligation induces production of D-3 phosphoinositides and tyrosine phosphorylation.
Flow Cytometry, SDS-Page, Immunofluorescence
0.2 mg/mL
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml
Purified
CD7
Chronic Lymphoid Leukemia cells (CLL)
40 kDa
0.02 mg
Cytokine Research, Signal Transduction
Primary
924
Human, Porcine, Guinea Pig
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only