Learn More
CD80 Mouse anti-Human, APC-R700, Clone: L307.4, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 565157
Description
The L307 monoclonal antibody specifically binds to B7/BB1, a 60 kDa transmembrane glycoprotein that was clustered as CD80 in the Fifth International Workshop on Human Leukocyte Differentiation Antigens. CD80, a member of the Ig supergene family, is expressed on activated B cells, T cells, macrophages, and dendritic cells. It is the ligand for two molecules expressed on T cells, CD28 and CD152 (CTLA-4). CD80 is also expressed on activated CD4-positive and CD8-positive T cells, appearing late after activation suggesting that activated T cells may be capable of autocrine costimulation via the CD28 activation pathway. The binding of CD28 by anti-CD28 or by CD80 results in T-cell activation and a signal for IL-2 production.
This antibody was conjugated to BD Horizon APC-R700, which has been developed exclusively by BD Biosciences as a better alternative to Alexa Fluor™ 700. APC-R700 excites and emits at similar wavelengths to Alexa Fluor™ 700 yet exhibits significantly improved brightness. This dye can be excited by the red laser and detected with the same filter set as Alexa Fluor™ (eg, 730/45-nm filter).
Specifications
CD80 | |
Monoclonal | |
APC-R700 | |
Aqueous buffered solution containing BSA, protein stabilizer, glycerol, and ≤0.09% sodium azide. | |
Human B7-transfected L cells | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
L307.4 | |
B7.1; B7-1; Activation B7-1 antigen; B7; BB1; CD28LG; CD28LG1; LAB7; L307 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.