Learn More
Cd8b, Mouse, Clone: 341, Abnova™
Mouse monoclonal antibody raised against Cd8b.
Manufacturer:  Abnova Corporation MAB10162
Catalog No. 89-108-410
Add to cart
Description
Description
Specifications
Specifications
Cd8b
341
Mouse monoclonal antibody raised against Cd8b.
In PBS, pH 7.4 (0.09% sodium azide)
Liquid
Cd8b
Cd8b positive Wistar rat splenic T cell hybridomas.
Protein A purification
RUO
Primary
24931
Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Unconjugated
Flow Cytometry (1.5 ug/ml) The optimal working dilution should be determined by the end user.
Cd8b
Cd8b1
Mouse
IgG1
100 ug
Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Rat