Learn More
Mouse anti-CD90/Thy1, Clone: 5E10, Cy5.5, PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP14284725TESTS
 View more versions of this product
Catalog No. NBP14284725
Add to cart
Description
Description
CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD90/Thy1
50000000000
PerCP-Cyanine5.5
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
Affinity Purified
THY1
The immunogen for this antibody was CD90.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Flow Cytometry
0.2 mg/mL
Flow Cytometry 1:10-1:1000
P04216
CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Mouse
IgG1 Kappa
25 Tests
Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primary
7070