Learn More
Mouse anti-CD90/Thy1, Clone: F15-42-1, Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP2333160.1MG
 View more versions of this product
Catalog No. NBP23331601
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Human, Primate samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional, CyTOF-ready.
Specifications
Specifications
CD90/Thy1
F15-42-1
Unconjugated
10mM PBS - BSA free with No Preservative
Purified
THY1
Purified human brain Thy1.
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
CD90 (F15-42-1)
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Functional Assay, CyTOF
1.0 mg/mL
Flow Cytometry 0.5-1.0ug/million cells, ELISA 1-5ug/ml for coating, Immunocytochemistry/Immunofluorescence 0.5ug-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Functional, CyTOF-ready
P04216
CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Mouse
IgG1 Kappa
0.1 mg
Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primary
7070
Human, Primate