Learn More
CD92 Mouse anti-Human, Alexa Fluor 647, Clone: VIM15, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565316
Catalog No. BDB565316
Add to cart
Description
Description
The VIM15 monoclonal antibody specifically binds to CD92 which is also known as Solute carrier family 44 member 1 (SLC44A1), or Choline transporter-like protein 1 (CHTL1). CD92 is a multipass membrane glycoprotein that belongs to the choline transporter-like family. It contains a tyrosine-based inhibitory motif (ITIM) in its cytoplasmic C-terminus. CD92 appears to be involved in choline transport for cell membrane phospholipid synthesis and in regulatory cell-signaling pathways. CD92 is strongly expressed by monocytes and at lower levels by granulocytes and some dendritic cells. It is generally expressed at low levels by B cells, endothelial cells, epithelial cells, and subsets of T cells, or NK cells. The VIM15 antibody has been shown to augment lipopolysaccharide-induced IL-10 production by monocyte-derived dendritic cells (Mo-DC). When cultured with ionomycin or calcium ionophore, maturing Mo-DC showed downregulation of CD92 expression.
Specifications
Specifications
CD92
VIM15
SLC44A1; Choline transporter-like protein 1; CHTL1; CTL1; RP11-287A8.1; p70
Affinity Purified
Human Myeloid Cell Line MV4-11
Affinity Chromatography
RUO
Primary
Human
Flow Cytometry
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgG1, κ
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal